Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601066 MIM: 603593 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OXA1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
OXA1L - OXA1L, mitochondrial inner membrane protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A7 - solute carrier family 7 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126105.2 | 1361 | Silent Mutation | TTA,TTG | L,L 373 | NP_001119577.1 | |
NM_001126106.2 | 1361 | Silent Mutation | TTA,TTG | L,L 373 | NP_001119578.1 | |
XM_006720302.1 | 1361 | Silent Mutation | TTA,TTG | L,L 373 | XP_006720365.1 | |
XM_011537298.2 | 1361 | Silent Mutation | TTA,TTG | L,L 373 | XP_011535600.1 | |
XM_011537299.1 | 1361 | Silent Mutation | TTA,TTG | L,L 373 | XP_011535601.1 |
Set Membership: |
DME Validated Inventoried |