Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 611510 MIM: 605813 MIM: 609119 | |||||||||||||||||||||||||||||
Literature Links: |
CENPT PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CENPT - centromere protein T | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025082.3 | 1135 | Intron | NP_079358.3 | |||
XM_006721277.2 | 1135 | Intron | XP_006721340.1 | |||
XM_011523344.1 | 1135 | Intron | XP_011521646.1 | |||
XM_011523345.1 | 1135 | Intron | XP_011521647.1 | |||
XM_017023712.1 | 1135 | Intron | XP_016879201.1 | |||
XM_017023713.1 | 1135 | Intron | XP_016879202.1 | |||
XM_017023714.1 | 1135 | Intron | XP_016879203.1 | |||
XM_017023715.1 | 1135 | Intron | XP_016879204.1 | |||
XM_017023716.1 | 1135 | Intron | XP_016879205.1 | |||
XM_017023717.1 | 1135 | Intron | XP_016879206.1 |
NUTF2 - nuclear transport factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THAP11 - THAP domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020457.2 | 1135 | Missense Mutation | CGT,CTT | R,L 297 | NP_065190.2 |