Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
80 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606891 MIM: 609251 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCRLA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FCRLA - Fc receptor like A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FCRLB - Fc receptor like B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002901.3 | 100 | UTR 5 | NP_001002901.1 | |||
NM_001288829.1 | 100 | Intron | NP_001275758.1 | |||
NM_001288830.1 | 100 | Intron | NP_001275759.1 | |||
NM_001288831.1 | 100 | Intron | NP_001275760.1 | |||
NM_001288832.1 | 100 | Intron | NP_001275761.1 | |||
NM_001320241.1 | 100 | Intron | NP_001307170.1 |
Set Membership: |
HapMap |