Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603944 | ||||||||||||||||||||
Literature Links: |
STX6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STX6 - syntaxin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286210.1 | Intron | NP_001273139.1 | ||||
NM_005819.5 | Intron | NP_005810.1 | ||||
XM_011509058.1 | Intron | XP_011507360.1 | ||||
XM_011509059.1 | Intron | XP_011507361.1 | ||||
XM_017000006.1 | Intron | XP_016855495.1 | ||||
XM_017000007.1 | Intron | XP_016855496.1 |