Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATAAATAGAAAAATTTTCTTCC[A/G]ACTTTGAAGACTAAAATATCTGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616337 MIM: 608126 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOCS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SOCS4 - suppressor of cytokine signaling 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080867.2 | Intron | NP_543143.1 | ||||
NM_199421.1 | Intron | NP_955453.1 | ||||
XM_011536425.1 | Intron | XP_011534727.1 | ||||
XM_011536426.1 | Intron | XP_011534728.1 |
WDHD1 - WD repeat and HMG-box DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |