Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605778 MIM: 601182 | ||||||||||||||||||||
Literature Links: |
NIF3L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NIF3L1 - NGG1 interacting factor 3 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORC2 - origin recognition complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006190.4 | 3126 | UTR 3 | NP_006181.1 | |||
XM_006712555.3 | 3126 | UTR 3 | XP_006712618.1 | |||
XM_011511252.2 | 3126 | UTR 3 | XP_011509554.1 | |||
XM_011511253.2 | 3126 | UTR 3 | XP_011509555.1 |