Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614560 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAU2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAU2 - MAU2 sister chromatid cohesion factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015329.3 | Intron | NP_056144.3 | ||||
XM_005259837.3 | Intron | XP_005259894.2 | ||||
XM_005259838.3 | Intron | XP_005259895.3 | ||||
XM_006722711.3 | Intron | XP_006722774.2 | ||||
XM_011527844.2 | Intron | XP_011526146.1 | ||||
XM_017026539.1 | Intron | XP_016882028.1 |