Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGCCGATTCCCAGAAACGCTCC[A/G]TTGGAGAATGCCGTTGAGATGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606091 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100129083 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC100129083 - uncharacterized LOC100129083 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIGLEC10 - sialic acid binding Ig like lectin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171156.1 | 1413 | Silent Mutation | NP_001164627.1 | |||
NM_001171157.1 | 1413 | Intron | NP_001164628.1 | |||
NM_001171158.1 | 1413 | Silent Mutation | NP_001164629.1 | |||
NM_001171159.1 | 1413 | Silent Mutation | NP_001164630.1 | |||
NM_001171161.1 | 1413 | Silent Mutation | NP_001164632.1 | |||
NM_001322105.1 | 1413 | Silent Mutation | NP_001309034.1 | |||
NM_033130.4 | 1413 | Silent Mutation | NP_149121.2 | |||
XM_005259366.4 | 1413 | Silent Mutation | XP_005259423.1 | |||
XM_005259367.3 | 1413 | Silent Mutation | XP_005259424.1 | |||
XM_011527439.1 | 1413 | Silent Mutation | XP_011525741.1 |
Set Membership: |
HapMap Validated |