Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604495 MIM: 604480 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCER2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CCER2 - coiled-coil glutamate rich protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFKBIB - NFKB inhibitor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243116.1 | Intron | NP_001230045.1 | ||||
NM_002503.4 | Intron | NP_002494.2 | ||||
XM_006723226.2 | Intron | XP_006723289.2 | ||||
XM_006723227.3 | Intron | XP_006723290.3 |
SIRT2 - sirtuin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193286.1 | Intron | NP_001180215.1 | ||||
NM_012237.3 | Intron | NP_036369.2 | ||||
NM_030593.2 | Intron | NP_085096.1 | ||||
XM_006723111.1 | Intron | XP_006723174.1 | ||||
XM_011526654.1 | Intron | XP_011524956.1 | ||||
XM_011526655.1 | Intron | XP_011524957.1 | ||||
XM_017026500.1 | Intron | XP_016881989.1 |