Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCACAAGAGGGCTCCGAGCAGA[A/G]GAGGGATATGATCTGACTCAGGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607419 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GEMIN7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GEMIN7 - gem nuclear organelle associated protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007269.1 | Intron | NP_001007270.1 | ||||
NM_001007270.1 | Intron | NP_001007271.1 | ||||
NM_001319054.1 | Intron | NP_001305983.1 | ||||
NM_001319055.1 | Intron | NP_001305984.1 | ||||
NM_024707.2 | Intron | NP_078983.1 | ||||
XM_005259262.3 | Intron | XP_005259319.1 | ||||
XM_005259263.4 | Intron | XP_005259320.1 | ||||
XM_017027312.1 | Intron | XP_016882801.1 |
LOC105372419 - uncharacterized LOC105372419 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R37 - protein phosphatase 1 regulatory subunit 37 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |