Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCACACTCCCCAACCTAAGACC[C/T]TGAGGGTCCCAGGCAGCACAGGTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607779 | ||||||||||||||||||||
Literature Links: |
AUNIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AUNIP - aurora kinase A and ninein interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAQR7 - progestin and adipoQ receptor family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178422.5 | 2675 | UTR 3 | NP_848509.1 | |||
XM_005245745.3 | 2675 | UTR 3 | XP_005245802.1 | |||
XM_005245746.3 | 2675 | UTR 3 | XP_005245803.1 | |||
XM_011540861.1 | 2675 | UTR 3 | XP_011539163.1 | |||
XM_011540862.2 | 2675 | UTR 3 | XP_011539164.1 |