Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611853 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL48 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MRPL48 - mitochondrial ribosomal protein L48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318498.1 | Intron | NP_001305427.1 | ||||
NM_001318499.1 | Intron | NP_001305428.1 | ||||
NM_001318500.1 | Intron | NP_001305429.1 | ||||
NM_016055.5 | Intron | NP_057139.1 | ||||
XM_017017919.1 | Intron | XP_016873408.1 |
Set Membership: |
HapMap |