Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 193040 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP37 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
USP37 - ubiquitin specific peptidase 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020935.2 | 7913 | UTR 3 | NP_065986.2 | |||
XM_005246720.2 | 7913 | UTR 3 | XP_005246777.1 | |||
XM_005246721.3 | 7913 | UTR 3 | XP_005246778.1 | |||
XM_005246722.3 | 7913 | UTR 3 | XP_005246779.1 | |||
XM_005246724.1 | 7913 | UTR 3 | XP_005246781.1 | |||
XM_011511538.2 | 7913 | UTR 3 | XP_011509840.1 | |||
XM_011511541.2 | 7913 | Intron | XP_011509843.1 | |||
XM_017004577.1 | 7913 | UTR 3 | XP_016860066.1 |
VIL1 - villin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007127.2 | 7913 | Intron | NP_009058.2 |
Set Membership: |
HapMap |