Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601958 MIM: 612172 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CACNB3 - calcium voltage-gated channel auxiliary subunit beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000725.3 | Intron | NP_000716.2 | ||||
NM_001206915.1 | Intron | NP_001193844.1 | ||||
NM_001206916.1 | Intron | NP_001193845.1 | ||||
NM_001206917.1 | Intron | NP_001193846.1 | ||||
XM_011538727.2 | Intron | XP_011537029.1 | ||||
XM_011538728.2 | Intron | XP_011537030.1 | ||||
XM_017019957.1 | Intron | XP_016875446.1 | ||||
XM_017019958.1 | Intron | XP_016875447.1 | ||||
XM_017019959.1 | Intron | XP_016875448.1 |
DDX23 - DEAD-box helicase 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004818.2 | Intron | NP_004809.2 |
Set Membership: |
HapMap Validated |