Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600016 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNTN1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CNTN1 - contactin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256063.1 | Intron | NP_001242992.1 | ||||
NM_001256064.1 | Intron | NP_001242993.1 | ||||
NM_001843.3 | Intron | NP_001834.2 | ||||
NM_175038.2 | Intron | NP_778203.1 | ||||
XM_005268651.2 | Intron | XP_005268708.1 | ||||
XM_006719241.2 | Intron | XP_006719304.1 | ||||
XM_011537926.2 | Intron | XP_011536228.1 | ||||
XM_011537927.2 | Intron | XP_011536229.1 | ||||
XM_017018821.1 | Intron | XP_016874310.1 | ||||
XM_017018822.1 | Intron | XP_016874311.1 | ||||
XM_017018823.1 | Intron | XP_016874312.1 | ||||
XM_017018824.1 | Intron | XP_016874313.1 | ||||
XM_017018825.1 | Intron | XP_016874314.1 | ||||
XM_017018826.1 | Intron | XP_016874315.1 | ||||
XM_017018827.1 | Intron | XP_016874316.1 |