Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606608 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
YAP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
YAP1 - Yes associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130145.2 | Intron | NP_001123617.1 | ||||
NM_001195044.1 | Intron | NP_001181973.1 | ||||
NM_001195045.1 | Intron | NP_001181974.1 | ||||
NM_001282097.1 | Intron | NP_001269026.1 | ||||
NM_001282098.1 | Intron | NP_001269027.1 | ||||
NM_001282099.1 | Intron | NP_001269028.1 | ||||
NM_001282100.1 | Intron | NP_001269029.1 | ||||
NM_001282101.1 | Intron | NP_001269030.1 | ||||
NM_006106.4 | Intron | NP_006097.2 | ||||
XM_005271378.3 | Intron | XP_005271435.1 | ||||
XM_005271380.3 | Intron | XP_005271437.1 | ||||
XM_005271381.3 | Intron | XP_005271438.1 | ||||
XM_005271383.3 | Intron | XP_005271440.1 | ||||
XM_011542555.2 | Intron | XP_011540857.1 | ||||
XM_011542556.2 | Intron | XP_011540858.1 | ||||
XM_017017093.1 | Intron | XP_016872582.1 |
Set Membership: |
HapMap |