Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120355 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMP27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MMP27 - matrix metallopeptidase 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP8 - matrix metallopeptidase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304441.1 | 1639 | UTR 3 | NP_001291370.1 | |||
NM_001304442.1 | 1639 | UTR 3 | NP_001291371.1 | |||
NM_002424.2 | 1639 | UTR 3 | NP_002415.1 | |||
XM_011542834.2 | 1639 | UTR 3 | XP_011541136.1 | |||
XM_011542835.2 | 1639 | UTR 3 | XP_011541137.1 | |||
XM_011542836.2 | 1639 | UTR 3 | XP_011541138.1 | |||
XM_017017771.1 | 1639 | UTR 3 | XP_016873260.1 |