Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613165 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CANT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CANT1 - calcium activated nucleotidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159772.1 | 3064 | UTR 3 | NP_001153244.1 | |||
NM_001159773.1 | 3064 | UTR 3 | NP_001153245.1 | |||
NM_138793.3 | 3064 | UTR 3 | NP_620148.1 | |||
XM_005257021.1 | 3064 | UTR 3 | XP_005257078.1 | |||
XM_005257022.1 | 3064 | UTR 3 | XP_005257079.1 | |||
XM_006721683.1 | 3064 | UTR 3 | XP_006721746.1 | |||
XM_011524291.1 | 3064 | UTR 3 | XP_011522593.1 | |||
XM_011524293.1 | 3064 | UTR 3 | XP_011522595.1 | |||
XM_011524294.1 | 3064 | UTR 3 | XP_011522596.1 | |||
XM_011524295.2 | 3064 | UTR 3 | XP_011522597.1 |
Set Membership: |
HapMap |