Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTTCATGGTCTGATTTATTGGT[A/G]GTGAATACACAGGGGCAGGCCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 131195 MIM: 136510 | ||||||||||||||||||||
Literature Links: |
ENG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENG - endoglin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000118.3 | 2776 | UTR 3 | NP_000109.1 | |||
NM_001114753.2 | 2776 | UTR 3 | NP_001108225.1 | |||
NM_001278138.1 | 2776 | UTR 3 | NP_001265067.1 |
FPGS - folylpolyglutamate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018078.2 | 2776 | Intron | NP_001018088.1 | |||
NM_001288803.1 | 2776 | Intron | NP_001275732.1 | |||
NM_004957.5 | 2776 | Intron | NP_004948.4 | |||
XM_005251864.3 | 2776 | Intron | XP_005251921.1 | |||
XM_011518439.2 | 2776 | Intron | XP_011516741.1 | |||
XM_017014565.1 | 2776 | Intron | XP_016870054.1 | |||
XM_017014566.1 | 2776 | Intron | XP_016870055.1 |
LOC102723566 - uncharacterized LOC102723566 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |