Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609720 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRB2 - crumbs 2, cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173689.6 | 540 | Intron | NP_775960.4 | |||
XM_005251934.2 | 540 | Intron | XP_005251991.1 | |||
XM_011518556.2 | 540 | Intron | XP_011516858.1 | |||
XM_011518557.2 | 540 | UTR 5 | XP_011516859.1 | |||
XM_011518558.2 | 540 | UTR 5 | XP_011516860.1 |
Set Membership: |
HapMap |