Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609147 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIC8B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RIC8B - RIC8 guanine nucleotide exchange factor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005268998.3 | Intron | XP_005269055.1 | ||||
XM_011538526.2 | Intron | XP_011536828.1 | ||||
XM_011538527.2 | Intron | XP_011536829.1 | ||||
XM_011538528.2 | Intron | XP_011536830.1 | ||||
XM_011538529.2 | Intron | XP_011536831.1 | ||||
XM_017019548.1 | Intron | XP_016875037.1 | ||||
XM_017019549.1 | Intron | XP_016875038.1 |