Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609264 MIM: 613687 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP37 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NUP37 - nucleoporin 37 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARPBP - PARP1 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319988.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | NP_001306917.1 | |
NM_001319993.1 | 200 | UTR 5 | NP_001306922.1 | |||
NM_001319994.1 | 200 | UTR 5 | NP_001306923.1 | |||
NM_001319995.1 | 200 | UTR 5 | NP_001306924.1 | |||
NM_001319996.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | NP_001306925.1 | |
NM_017915.4 | 200 | Silent Mutation | ACC,ACT | T,T 29 | NP_060385.3 | |
XM_011538509.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | XP_011536811.1 | |
XM_011538514.2 | 200 | Intron | XP_011536816.1 | |||
XM_011538515.1 | 200 | Intron | XP_011536817.1 | |||
XM_017019534.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | XP_016875023.1 | |
XM_017019535.1 | 200 | Intron | XP_016875024.1 | |||
XM_017019536.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | XP_016875025.1 | |
XM_017019537.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | XP_016875026.1 | |
XM_017019538.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | XP_016875027.1 | |
XM_017019539.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | XP_016875028.1 | |
XM_017019540.1 | 200 | UTR 5 | XP_016875029.1 | |||
XM_017019541.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | XP_016875030.1 | |
XM_017019542.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | XP_016875031.1 | |
XM_017019543.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | XP_016875032.1 | |
XM_017019544.1 | 200 | Silent Mutation | ACC,ACT | T,T 29 | XP_016875033.1 |