Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGGCTGCCTCTGGGGTCGGCAT[A/G]GGAGCAGGCCTTCCCTTGCAGCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614297 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C19orf12 - chromosome 19 open reading frame 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031726.3 | 3763 | UTR 3 | NP_001026896.2 | |||
NM_001256046.1 | 3763 | UTR 3 | NP_001242975.1 | |||
NM_001256047.1 | 3763 | UTR 3 | NP_001242976.1 | |||
NM_001282929.1 | 3763 | UTR 3 | NP_001269858.1 | |||
NM_001282930.1 | 3763 | UTR 3 | NP_001269859.1 | |||
NM_001282931.1 | 3763 | UTR 3 | NP_001269860.1 | |||
NM_031448.4 | 3763 | UTR 3 | NP_113636.2 |
Set Membership: |
HapMap |