Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCTTAAAAATAAACATGGACTG[C/G]ATTAGCAGAGCAGAGCAGGCGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC284395 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC284395 - uncharacterized LOC284395 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VSTM2B - V-set and transmembrane domain containing 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146339.1 | Intron | NP_001139811.1 | ||||
XM_005258856.3 | Intron | XP_005258913.1 | ||||
XM_011526902.2 | Intron | XP_011525204.1 | ||||
XM_011526903.2 | Intron | XP_011525205.1 | ||||
XM_011526904.2 | Intron | XP_011525206.1 | ||||
XM_011526905.2 | Intron | XP_011525207.1 |