Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147440 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IGF1 - insulin like growth factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000618.4 | 3244 | UTR 3 | NP_000609.1 | |||
NM_001111283.2 | 3244 | UTR 3 | NP_001104753.1 | |||
NM_001111284.1 | 3244 | UTR 3 | NP_001104754.1 | |||
XM_017019259.1 | 3244 | Intron | XP_016874748.1 | |||
XM_017019260.1 | 3244 | Intron | XP_016874749.1 | |||
XM_017019261.1 | 3244 | Intron | XP_016874750.1 | |||
XM_017019262.1 | 3244 | UTR 3 | XP_016874751.1 | |||
XM_017019263.1 | 3244 | UTR 3 | XP_016874752.1 |
Set Membership: |
HapMap |