Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606920 MIM: 604396 | ||||||||||||||||||||
Literature Links: |
CERS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CERS2 - ceramide synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022075.4 | Intron | NP_071358.1 | ||||
NM_181746.3 | Intron | NP_859530.1 | ||||
XM_011509451.2 | Intron | XP_011507753.1 | ||||
XM_011509452.2 | Intron | XP_011507754.1 |
SETDB1 - SET domain bifurcated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |