Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602932 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMAD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMAD7 - SMAD family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190821.1 | 479 | Silent Mutation | CTA,CTG | L,L 297 | NP_001177750.1 | |
NM_001190822.1 | 479 | Silent Mutation | CTA,CTG | L,L 83 | NP_001177751.1 | |
NM_001190823.1 | 479 | Silent Mutation | CTA,CTG | L,L 110 | NP_001177752.1 | |
NM_005904.3 | 479 | Silent Mutation | CTA,CTG | L,L 298 | NP_005895.1 |