Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAATCCCGGGGGTCTCCCGCGTC[C/T]CCATACCCCCACCCCGGCCTCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612665 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEX101 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TEX101 - testis expressed 101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130011.1 | Intron | NP_001123483.1 | ||||
NM_031451.4 | Intron | NP_113639.4 | ||||
XM_005259303.3 | Intron | XP_005259360.2 |