Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614659 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMER2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AMER2 - APC membrane recruitment protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152704.3 | 6056 | UTR 3 | NP_689917.2 | |||
NM_199138.2 | 6056 | UTR 3 | NP_954589.1 | |||
XM_005266279.3 | 6056 | UTR 3 | XP_005266336.1 | |||
XM_005266280.3 | 6056 | UTR 3 | XP_005266337.1 | |||
XM_017020415.1 | 6056 | UTR 3 | XP_016875904.1 |
LINC00463 - long intergenic non-protein coding RNA 463 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |