Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATTCATCTGCCCTCGGGCATAG[A/G]GGTGGAGGCTGCCATGCCCAGGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604638 MIM: 608839 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTN4 - actinin alpha 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CAPN12 - calpain 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144691.4 | Intron | NP_653292.2 | ||||
XM_017026355.1 | Intron | XP_016881844.1 | ||||
XM_017026356.1 | Intron | XP_016881845.1 | ||||
XM_017026357.1 | Intron | XP_016881846.1 | ||||
XM_017026358.1 | Intron | XP_016881847.1 | ||||
XM_017026359.1 | Intron | XP_016881848.1 | ||||
XM_017026360.1 | Intron | XP_016881849.1 | ||||
XM_017026361.1 | Intron | XP_016881850.1 | ||||
XM_017026362.1 | Intron | XP_016881851.1 | ||||
XM_017026363.1 | Intron | XP_016881852.1 | ||||
XM_017026364.1 | Intron | XP_016881853.1 | ||||
XM_017026365.1 | Intron | XP_016881854.1 |