Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTCTTCCGCCTGTCACCACTT[C/G]TCTCTGGGCCCTGCCTCTGAACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614545 MIM: 615324 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASPDH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASPDH - aspartate dehydrogenase domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EMC10 - ER membrane protein complex subunit 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
JOSD2 - Josephin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270639.1 | Intron | NP_001257568.1 | ||||
NM_001270640.1 | Intron | NP_001257569.1 | ||||
NM_001270641.1 | Intron | NP_001257570.1 | ||||
NM_001270686.1 | Intron | NP_001257615.1 | ||||
NM_138334.3 | Intron | NP_612207.1 | ||||
XM_011526434.2 | Intron | XP_011524736.1 |
LRRC4B - leucine rich repeat containing 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |