Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCAAGCCAATAAAATAGAGCAC[A/G]TATCTCAGCAGAACTGCAGACAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF561 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF561 - zinc finger protein 561 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152289.2 | 4225 | UTR 3 | NP_689502.2 | |||
XM_005260150.2 | 4225 | UTR 3 | XP_005260207.1 | |||
XM_005260151.2 | 4225 | UTR 3 | XP_005260208.1 | |||
XM_006722938.2 | 4225 | UTR 3 | XP_006723001.1 | |||
XM_011528433.2 | 4225 | UTR 3 | XP_011526735.1 | |||
XM_011528434.2 | 4225 | UTR 3 | XP_011526736.1 | |||
XM_011528436.2 | 4225 | UTR 3 | XP_011526738.1 | |||
XM_011528437.2 | 4225 | UTR 3 | XP_011526739.1 | |||
XM_017027478.1 | 4225 | UTR 3 | XP_016882967.1 | |||
XM_017027479.1 | 4225 | UTR 3 | XP_016882968.1 | |||
XM_017027480.1 | 4225 | UTR 3 | XP_016882969.1 | |||
XM_017027481.1 | 4225 | UTR 3 | XP_016882970.1 | |||
XM_017027482.1 | 4225 | UTR 3 | XP_016882971.1 |