Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606100 MIM: 611811 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADGRE2 - adhesion G protein-coupled receptor E2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271052.1 | 1808 | Intron | NP_001257981.1 | |||
NM_013447.3 | 1808 | Intron | NP_038475.2 | |||
XM_011527948.2 | 1808 | Intron | XP_011526250.1 | |||
XM_011527949.2 | 1808 | Intron | XP_011526251.1 | |||
XM_011527951.2 | 1808 | Intron | XP_011526253.1 | |||
XM_011527952.2 | 1808 | Intron | XP_011526254.1 | |||
XM_011527953.2 | 1808 | Intron | XP_011526255.1 | |||
XM_011527954.2 | 1808 | Intron | XP_011526256.1 | |||
XM_011527955.2 | 1808 | Intron | XP_011526257.1 | |||
XM_017026726.1 | 1808 | Intron | XP_016882215.1 | |||
XM_017026727.1 | 1808 | Intron | XP_016882216.1 |
ZNF333 - zinc finger protein 333 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300912.1 | 1808 | UTR 3 | NP_001287841.1 | |||
NM_032433.2 | 1808 | Intron | NP_115809.1 | |||
XM_005260098.3 | 1808 | Intron | XP_005260155.1 | |||
XM_006722924.2 | 1808 | Intron | XP_006722987.1 | |||
XM_011528362.2 | 1808 | Intron | XP_011526664.1 | |||
XM_011528363.2 | 1808 | Intron | XP_011526665.1 | |||
XM_011528365.2 | 1808 | Intron | XP_011526667.1 | |||
XM_011528366.2 | 1808 | Intron | XP_011526668.1 | |||
XM_011528367.2 | 1808 | Intron | XP_011526669.1 | |||
XM_011528368.2 | 1808 | Intron | XP_011526670.1 | |||
XM_011528369.2 | 1808 | Intron | XP_011526671.1 | |||
XM_011528371.2 | 1808 | Intron | XP_011526673.1 | |||
XM_011528372.2 | 1808 | Intron | XP_011526674.1 | |||
XM_017027367.1 | 1808 | Intron | XP_016882856.1 | |||
XM_017027368.1 | 1808 | Intron | XP_016882857.1 | |||
XM_017027369.1 | 1808 | Intron | XP_016882858.1 | |||
XM_017027370.1 | 1808 | Intron | XP_016882859.1 | |||
XM_017027371.1 | 1808 | Intron | XP_016882860.1 |
Set Membership: |
HapMap Validated |