Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAAAGTGCATGAGGTTACCAGGT[A/C]CTAGGTTCCAGGGTGCTCCTCCTAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611998 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREB3L3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CREB3L3 - cAMP responsive element binding protein 3 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271995.1 | Intron | NP_001258924.1 | ||||
NM_001271996.1 | Intron | NP_001258925.1 | ||||
NM_001271997.1 | Intron | NP_001258926.1 | ||||
NM_032607.2 | Intron | NP_115996.1 |