Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCATTGGCCCTGGGACGCTGTC[A/G]GAGATTCAGATGTGAACCCTCCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608229 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf57 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C19orf57 - chromosome 19 open reading frame 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024323.3 | 2491 | Intron | NP_077299.3 | |||
XM_006722885.3 | 2491 | Intron | XP_006722948.2 | |||
XM_011528286.2 | 2491 | UTR 3 | XP_011526588.1 | |||
XM_011528287.2 | 2491 | UTR 3 | XP_011526589.1 | |||
XM_017027300.1 | 2491 | Intron | XP_016882789.1 | |||
XM_017027301.1 | 2491 | Intron | XP_016882790.1 |
MIR181D - microRNA 181d | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NANOS3 - nanos C2HC-type zinc finger 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |