Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612419 MIM: 609435 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CILP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CILP2 - cartilage intermediate layer protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFA13 - NADH:ubiquinone oxidoreductase subunit A13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YJEFN3 - YjeF N-terminal domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190328.1 | 292 | Intron | NP_001177257.1 | |||
NM_198537.3 | 292 | Intron | NP_940939.2 | |||
XM_005259903.4 | 292 | Intron | XP_005259960.1 | |||
XM_011527996.2 | 292 | Intron | XP_011526298.1 | |||
XM_011527997.1 | 292 | UTR 5 | XP_011526299.1 | |||
XM_011527998.2 | 292 | Intron | XP_011526300.1 | |||
XM_011527999.1 | 292 | Intron | XP_011526301.1 | |||
XM_017026770.1 | 292 | Silent Mutation | CTA,TTA | L,L 11 | XP_016882259.1 | |
XM_017026771.1 | 292 | UTR 5 | XP_016882260.1 |
Set Membership: |
HapMap Validated |