Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATGTTCAGTCCATGTGTCCTTA[A/C]AAGTGAAGCAAGTTTCTTCTAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609486 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EAPP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EAPP - E2F associated phosphoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318916.1 | 762 | Intron | NP_001305845.1 | |||
NM_018453.3 | 762 | Intron | NP_060923.2 | |||
XM_011536958.1 | 762 | Intron | XP_011535260.1 | |||
XM_017021460.1 | 762 | Missense Mutation | TGT,TTT | C,F 218 | XP_016876949.1 |