Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603647 MIM: 616014 MIM: 607652 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCS1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BCS1L - BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF25 - ring finger protein 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022453.2 | Intron | NP_071898.2 | ||||
XM_017004695.1 | Intron | XP_016860184.1 |
STK36 - serine/threonine kinase 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243313.1 | Intron | NP_001230242.1 | ||||
NM_015690.4 | Intron | NP_056505.2 | ||||
XM_005246464.1 | Intron | XP_005246521.1 | ||||
XM_011510959.1 | Intron | XP_011509261.1 | ||||
XM_011510960.1 | Intron | XP_011509262.1 | ||||
XM_011510961.1 | Intron | XP_011509263.1 | ||||
XM_017003804.1 | Intron | XP_016859293.1 | ||||
XM_017003805.1 | Intron | XP_016859294.1 |
Set Membership: |
Validated |