Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGCTCACAGGCCCAGAGGGGAC[A/G]GAGAAGGGGTTACCTCCGTTCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603832 MIM: 606862 MIM: 606419 MIM: 609519 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NDUFA3 - NADH:ubiquinone oxidoreductase subunit A3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSCAR - osteoclast associated, immunoglobulin-like receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRPF31 - pre-mRNA processing factor 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TFPT - TCF3 fusion partner | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321792.1 | Intron | NP_001308721.1 | ||||
NM_013342.3 | Intron | NP_037474.1 | ||||
XM_005278261.1 | Intron | XP_005278318.1 |