Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606526 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLPH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MLPH - melanophilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042467.2 | Intron | NP_001035932.1 | ||||
NM_001281473.1 | Intron | NP_001268402.1 | ||||
NM_001281474.1 | Intron | NP_001268403.1 | ||||
NM_024101.6 | Intron | NP_077006.1 | ||||
XM_006712737.1 | Intron | XP_006712800.1 | ||||
XM_006712739.1 | Intron | XP_006712802.1 | ||||
XM_006712740.1 | Intron | XP_006712803.1 | ||||
XM_011511812.1 | Intron | XP_011510114.1 | ||||
XM_017004893.1 | Intron | XP_016860382.1 | ||||
XM_017004894.1 | Intron | XP_016860383.1 |
Set Membership: |
HapMap Validated |