Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613294 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAE1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SAE1 - SUMO1 activating enzyme subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145713.1 | Intron | NP_001139185.1 | ||||
NM_001145714.1 | Intron | NP_001139186.1 | ||||
NM_005500.2 | Intron | NP_005491.1 | ||||
XM_006722963.2 | Intron | XP_006723026.1 | ||||
XM_006722965.3 | Intron | XP_006723028.1 | ||||
XM_017026135.1 | Intron | XP_016881624.1 | ||||
XM_017026136.1 | Intron | XP_016881625.1 |
Set Membership: |
HapMap |