Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608525 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ING5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ING5 - inhibitor of growth family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005247047.2 | Intron | XP_005247104.1 | ||||
XM_017005097.1 | Intron | XP_016860586.1 | ||||
XM_017005098.1 | Intron | XP_016860587.1 | ||||
XM_017005099.1 | Intron | XP_016860588.1 | ||||
XM_017005100.1 | Intron | XP_016860589.1 | ||||
XM_017005101.1 | Intron | XP_016860590.1 | ||||
XM_017005102.1 | Intron | XP_016860591.1 |
Set Membership: |
HapMap |