Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGTGCCCAGCGGCAGCCCAAAC[A/T]GTGCCCAAACCCCCGATGAGACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611165 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPRX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DPRX - divergent-paired related homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012728.1 | Intron | NP_001012746.1 | ||||
XM_011527011.2 | Intron | XP_011525313.1 | ||||
XM_011527012.2 | Intron | XP_011525314.1 |
LOC284379 - solute carrier family 7 member 3 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |