Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605928 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ARFIP1 - ADP ribosylation factor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025593.2 | Intron | NP_001020764.1 | ||||
NM_001025595.2 | Intron | NP_001020766.1 | ||||
NM_001287431.1 | Intron | NP_001274360.1 | ||||
NM_001287432.1 | Intron | NP_001274361.1 | ||||
NM_001287433.1 | Intron | NP_001274362.1 | ||||
NM_014447.3 | Intron | NP_055262.1 | ||||
XM_011531868.2 | Intron | XP_011530170.1 | ||||
XM_011531869.2 | Intron | XP_011530171.1 | ||||
XM_011531870.2 | Intron | XP_011530172.1 | ||||
XM_011531871.2 | Intron | XP_011530173.1 | ||||
XM_011531872.1 | Intron | XP_011530174.1 |
TIGD4 - tigger transposable element derived 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |