Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 607950 MIM: 607237 | |||||||||||||||||||||||||||||
Literature Links: |
PRSS46 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
PRSS46 - protease, serine 46 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRSS50 - protease, serine 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013270.4 | Intron | NP_037402.1 |
TMIE - transmembrane inner ear | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |