Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCAGGGTTGAGGTGCTTGGGGT[A/G]CTGGGGGGGCCCGGGGCTTCACCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM83B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM83B - family with sequence similarity 83 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010872.2 | Intron | NP_001010872.1 | ||||
XM_006715022.3 | Intron | XP_006715085.1 | ||||
XM_011514394.2 | Intron | XP_011512696.1 | ||||
XM_011514395.2 | Intron | XP_011512697.1 | ||||
XM_017010478.1 | Intron | XP_016865967.1 |