Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCATTTTTGAAAGGTCTTGACTC[A/G]TGTTGTTGTTTTGACAGCAATTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611758 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTUD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
OTUD3 - OTU deubiquitinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015207.1 | Intron | NP_056022.1 | ||||
XM_005245792.3 | Intron | XP_005245849.1 | ||||
XM_005245793.3 | Intron | XP_005245850.1 | ||||
XM_005245794.3 | Intron | XP_005245851.1 |
Set Membership: |
HapMap Validated |