Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf141 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C6orf141 - chromosome 6 open reading frame 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145652.1 | 2434 | Intron | NP_001139124.1 | |||
XM_005248850.3 | 2434 | UTR 3 | XP_005248907.1 | |||
XM_005248852.2 | 2434 | UTR 3 | XP_005248909.1 | |||
XM_005248853.2 | 2434 | UTR 3 | XP_005248910.1 | |||
XM_006714997.2 | 2434 | UTR 3 | XP_006715060.1 | |||
XM_011514303.2 | 2434 | UTR 3 | XP_011512605.1 | |||
XM_017010294.1 | 2434 | UTR 3 | XP_016865783.1 | |||
XM_017010295.1 | 2434 | UTR 3 | XP_016865784.1 | |||
XM_017010296.1 | 2434 | UTR 3 | XP_016865785.1 | |||
XM_017010297.1 | 2434 | UTR 3 | XP_016865786.1 | |||
XM_017010298.1 | 2434 | UTR 3 | XP_016865787.1 |