Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGAGGACCATATACCTACCCA[C/T]AAGTGTCACAGTATTTGCAAAGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEL1L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEL1L3 - SEL1L family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297592.1 | Intron | NP_001284521.1 | ||||
NM_001297594.1 | Intron | NP_001284523.1 | ||||
NM_015187.4 | Intron | NP_056002.2 | ||||
XM_011513819.2 | Intron | XP_011512121.2 | ||||
XM_017007923.1 | Intron | XP_016863412.1 |