Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
63 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612494 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF10L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGEF10L - Rho guanine nucleotide exchange factor 10 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011722.2 | Intron | NP_001011722.2 | ||||
NM_001319837.1 | Intron | NP_001306766.1 | ||||
NM_001319838.1 | Intron | NP_001306767.1 | ||||
NM_001328124.1 | Intron | NP_001315053.1 | ||||
NM_018125.3 | Intron | NP_060595.3 | ||||
XM_005245923.2 | Intron | XP_005245980.2 | ||||
XM_005245925.2 | Intron | XP_005245982.2 | ||||
XM_005245927.2 | Intron | XP_005245984.2 | ||||
XM_005245929.2 | Intron | XP_005245986.2 | ||||
XM_006710728.1 | Intron | XP_006710791.1 | ||||
XM_006710729.1 | Intron | XP_006710792.1 | ||||
XM_006710731.1 | Intron | XP_006710794.1 | ||||
XM_011541691.2 | Intron | XP_011539993.1 | ||||
XM_011541692.2 | Intron | XP_011539994.1 | ||||
XM_011541693.2 | Intron | XP_011539995.1 | ||||
XM_011541695.2 | Intron | XP_011539997.1 | ||||
XM_011541696.2 | Intron | XP_011539998.1 | ||||
XM_017001617.1 | Intron | XP_016857106.1 | ||||
XM_017001618.1 | Intron | XP_016857107.1 | ||||
XM_017001619.1 | Intron | XP_016857108.1 | ||||
XM_017001620.1 | Intron | XP_016857109.1 | ||||
XM_017001621.1 | Intron | XP_016857110.1 | ||||
XM_017001622.1 | Intron | XP_016857111.1 |
Set Membership: |
HapMap |